Learn More
Thermo Scientific™ dI Phosphoramidite, standard grade, 20-400 finish bottle
Shop All Thermo Scientific ProductsDescription
dI CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyinosine (dI) for use in oligonucleotide synthesis. The problem of degeneracy of the genetic code can be assuaged by utilizing universal bases, such as deoxyinosine (dI), in an oligonucleotide. Its unique structure allows for it to pair with all four bases in various 'wobble' structures, with an overall preferential order of base-pairing: dI-dC > dI-dA > dI-dG = dI-dT. This product is supplied in a 30 mL amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.
Features:
• Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C40H47N6O7P
• Formula weight: 754.8 g/mol
• CAS number: 141684-35-7
• HPLC Purity: ≥98.0%; M-11 Impurity ≤0.01%
• 31P NMR Purity: ≥98%
Specifications
Specifications
| For Use With (Equipment) | Expedite Synthesizers |
| Product Type | dI CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Deoxyinosine (dI) |
| Shipping Condition | Room Temperature |
| Purity | 98% |
| Quantity | 1 g |
| Reagent Type | Phosphoramidite |
| Grade | Standard |
| Nucleic Acid Type | DNA |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.