Learn More
Bochem™ E-Poli Test Tube Stands
Specifications
Specifications
| Holds | Tubes (16 mm) |
| Material | 18/10 E-Poli |
| Positions | 24 |
| Diameter (Metric) Opening | 16 mm |
| Product Type | Test Tube Rack |
| Array | 4 x 6 |