missing translation for 'onlineSavingsMsg'
Learn More

di-Potassium hydrogen phosphate, BAKER, J.T. Baker™

Product Code. 10010470 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10010470 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10010470 Supplier JT Baker Supplier No. 0242.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Laboratory Reagent

Specifications

Melting Point 340°C
CAS 11-4-7758
pH 8.5 to 9.6 (5% Solution at 25°C)
Quantity 1 kg
Assay Percent Range 98.0 to 100.5%
Molecular Formula HK2O4P
MDL Number MFCD00011383
Synonym di-Potassium hydrogen orthophosphate , Dibasic Potassium Phosphate , Potassium phosphate dibasic , Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate) , Potassium hydrogen phosphate
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES [K+].[K+].OP([O-])([O-])=O
IUPAC Name dipotassium hydrogen phosphate
Molecular Weight (g/mol) 174.17
Identification Passes Test
Loss on Drying max. 1.0% (at 105°C)
Density 2.3 g/cm³ (20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.