missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate 0.25M Solution, Certified, for HPLC, Fisher Chemical

Product Code. 10503484 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10503484 500 mL Plastic Coated Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10503484 Supplier Fisher Chemical Supplier No. J/7650/PB08
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7558-79-4
pH 9.1
Physical Form Liquid
Quantity 500 mL
Packaging Plastic Coated Glass bottle
Molecular Formula HNa2O4P
MDL Number MFCD00003496
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES [Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium hydrogen phosphate
Molecular Weight (g/mol) 141.96
PubChem CID 24203
ChEBI CHEBI:34683
Formula Weight 141.98g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.