missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate Dihydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10756301 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10756301 1 kg Plastic powder jar 1kg
10122880 500 g Plastic powder jar 500g
10316440 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10756301 Supplier Fisher Chemical Supplier No. S/4440/60
  • ONLINE EXCLUSIVE PRICE
    81.55 €
    67.70 € / 1kg
    Save 13.85 €
    16% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
IUPAC Name disodium;hydrogen phosphate;dihydrate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 243°C
Color White
pH 8.7 to 9.4
Physical Form Solid
Quantity 1 kg
Formula Weight 177.99g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.