missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10656462 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10656462 1 kg 1kg
10458610 500 g 500g
10182730 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10656462 Supplier Fisher Chemical Supplier No. S/4360/60
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10039-32-4
Molecular Formula H25Na2O16P
Molecular Weight (g/mol) 358.137
MDL Number 149181
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
PubChem CID 61456
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Melting Point 35°C
pH 9.0 to 9.4
Physical Form Solid
Quantity 1 kg
Formula Weight 358.13g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.