missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate, for HPLC, Thermo Scientific
GREENER_CHOICE

Product Code. 30280943 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280943 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280943 Supplier Thermo Scientific Supplier No. T076500500
  • ONLINE EXCLUSIVE PRICE
    323.00 €
    39.10 € / 500mL
    Save 283.90 €
    87% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7558-79-4
Physical Form Liquid or Solid
Quantity 500 mL
Molecular Weight (g/mol) 141.96 g/mol
Grade HPLC
Concentration or Composition (by Analyte or Components) 0.25 M
Packaging Plastic Coated Glass Bottle
Chemical Name or Material Di-Sodium Hydrogen Orthophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.