missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium tetraborate decahydrate, BAKER, J.T. Baker™

Product Code. 10417850 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10417850 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10417850 Supplier JT Baker Supplier No. 0270.1000
  • ONLINE EXCLUSIVE PRICE
    66.96 €
    48.20 € / 1kg
    Save 18.76 €
    28% Off
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets Reagent Specifications for testing Ph. Eur. monographs

Specifications

Boiling Point 320°C (1013 hPa)
Melting Point 75°C
CAS 1303-96-4
pH 9.0 to 9.6
Quantity 1 kg
Assay Percent Range 99.0 to 103.0%
Molecular Formula B4H20Na2O17
MDL Number MFCD00149193
Synonym Sodium tetraborate decahydrate , Borax tetra
InChI Key OJPFLODMGUZMKD-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O
IUPAC Name disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
Molecular Weight (g/mol) 381.36
Identification Passes Test
Density 1.73 g/cm³ (20°C)
Show More Show Less
compliance-icons
Product Identifier
  • di-Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: di-Sodium tetraborate decahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.