missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Tetraborate Decahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10477650 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
250g
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10477650 500 g Plastic powder jar 500g
10020430 250 g Plastic powder jar 250g
10294540 1 kg Plastic powder jar 1kg
10192540 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10477650 Supplier Fisher Chemical Supplier No. S/7040/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Di-Sodium Tetraborate Decahydrate, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
MDL Number 149193

Specifications

Melting Point 75°C
Color White
pH 9
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 381.36g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.