missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl azodicarboxylate, 98%

Product Code. 11319983
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319983 25 g 25g
11309983 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319983 Supplier Thermo Scientific Alfa Aesar Supplier No. L00294.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 870-50-8 | C10H18N2O4 | 230.264 g/mol

Di-tert-butyl azodicarboxylate is a reagent used in the preparation of acyl hydrazinedicarboxylates. It is also used in the electrophilic amination of beta-keto esters catalyzed by an axially chiral guanidine. It serves as a precursor in an enantioselective synthesis of 3,6-dihyropyridazines employing organocatalysts such as L-proline or (S)-2-pyrrolidinyl tetrazole. It is also utilized in the asymmetric Friedel-Crafts amination through a chiral organocatalyst. Further, it acts as a reactant for preparation of hexapeptide key fragments through stereo selective selenocyclization/oxidative deselenylation reactions. In addition to this, it is employed as a starting material in the synthesis of pyrroloisoquinoline template through stereoselective N-acyliminium-mediated cyclization and enolate amination for preparation of peptidomimetic compounds and Barbier-type propargylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl azodicarboxylate is a reagent used in the preparation of acyl hydrazinedicarboxylates. It is also used in the electrophilic amination of beta-keto esters catalyzed by an axially chiral guanidine. It serves as a precursor in an enantioselective synthesis of 3,6-dihyropyridazines employing organocatalysts such as L-proline or (S)-2-pyrrolidinyl tetrazole. It is also utilized in the asymmetric Friedel-Crafts amination through a chiral organocatalyst. Further, it acts as a reactant for preparation of hexapeptide key fragments through stereo selective selenocyclization/oxidative deselenylation reactions. In addition to this, it is employed as a starting material in the synthesis of pyrroloisoquinoline template through stereoselective N-acyliminium-mediated cyclization and enolate amination for preparation of peptidomimetic compounds and Barbier-type propargylation reactions.

Solubility
Soluble in most organic solvents. Insoluble in water.

Notes
Light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-50-8
Molecular Formula C10H18N2O4
Molecular Weight (g/mol) 230.264
MDL Number MFCD00015001
InChI Key QKSQWQOAUQFORH-VAWYXSNFSA-N
Synonym di-tert-butyl azodicarboxylate, di-tert-butylazodicarboxylate, di-tert-butyl diazene-1,2-dicarboxylate, azodicarboxylic acid di-tert-butyl ester, di-tert-butyl azodicarboxylate dbad, pubchem17548, n-tert-butoxycarbonyl imino tert-butoxy formamide, tert-butyl tert-butyl oxycarbonyl diazenyl formate, n-tert-butoxy carbonyl imino tert-butoxy formamide, 1,2-diazenedicarboxylic acid, 1,2-bis 1,1-dimethylethyl ester
PubChem CID 6034084
IUPAC Name tert-butyl (NE)-N-[(2-methylpropan-2-yl)oxycarbonylimino]carbamate
SMILES CC(C)(C)OC(=O)N=NC(=O)OC(C)(C)C

Specifications

Melting Point 89°C to 92°C
Quantity 25 g
Beilstein 1911434
Sensitivity Light sensitive
Solubility Information Soluble in most organic solvents. Insoluble in water.
Formula Weight 230.27
Percent Purity 98%
Chemical Name or Material Di-tert-butyl azodicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.