missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl iminodicarboxylate, 97%

Product Code. 11915601 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11915601 100 g Glass Bottle 100g
10364122 5 g Glass Bottle 5g
10232722 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11915601 Supplier Thermo Scientific Acros Supplier No. 369661000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51779-32-9 | C10H19NO4 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51779-32-9
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00043309
InChI Key XCAQIUOFDMREBA-UHFFFAOYSA-N
Synonym di-tert-butyl iminodicarboxylate, di-t-butyl iminodicarboxylate, di-tert-butyl imidodicarbonate, n-boc-tert-butylcarbamate, tert-butyl iminodicarboxylate, di-tert-butyl iminodicarbonate, di-tert-butyl-iminodicarboxylate, iminodicarboxylic acid di-tert-butyl ester, tert-butyl n-tert-butoxy carbonyl carbamate, di tert-butyl imidodicarbonate
PubChem CID 279800
IUPAC Name tert-butyl N-[(2-methylpropan-2-yl)oxycarbonyl]carbamate
SMILES CC(C)(C)OC(=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 114.0°C to 117.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula [(CH3)3COCO]2NH
Fieser 01,210; 01,782; 02,303; 04,369; 05,500; 06,447; 07,277; 08,384; 09,352; 10,300; 11,393; 12,371; 13,9; 15,248; 16,268
Quantity 100 g
Formula Weight 217.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Di-tert-butyl iminodicarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Di-tert-butyl iminodicarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.