missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl malonate, 98%

Product Code. 10700482 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10700482 10 g 10g
10764204 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10700482 Supplier Thermo Scientific Acros Supplier No. 406620100
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 541-16-2 | C11H20O4 | 216.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-16-2
MDL Number MFCD00008810
InChI Key CLPHAYNBNTVRDI-UHFFFAOYSA-N
Synonym di-tert-butyl malonate, malonic acid di-tert-butyl ester, propanedioic acid, bis 1,1-dimethylethyl ester, di-t-butylmalonate, di-t-butyl malonate, unii-7e9xwt9380, 1,3-di-tert-butyl propanedioate, di tert-butyl malonate, tert-butyl 2-tert-butoxycarbonyl acetate, di-tert-butyl malonate, stab. with potassium carbonate
PubChem CID 68324
IUPAC Name ditert-butyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OC(C)(C)C

Specifications

Melting Point -7°C
Color Colorless to Yellow
Density 0.9660g/mL
Boiling Point 110°C to 111°C (22.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C11H20O4
Linear Formula CH2(CO2C(CH3)3)2
Refractive Index 1.4175 to 1.4195
Quantity 10 g
Beilstein 02, III, 1621
Fieser 01,210
Merck Index 15, 3041
Specific Gravity 0.966
Molecular Weight (g/mol) 216.28
Formula Weight 216.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Di-tert-butyl malonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.