missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl N,N-diethylphosphoramidite, 94%

Product Code. 11367169
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367169 1 g 1g
11377169 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367169 Supplier Thermo Scientific Alfa Aesar Supplier No. H27380.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 117924-33-1 | C12H28NO2P | 249.335 g/mol

A useful reagent for the efficient phosphorylative conversion of alcohols into their corresponding dibenzylphosphorotriesters. It is applied as a phosphitylating agent in water-soluble phosphate prodrugs of 1-propargyl-8-styrylxanthine derivatives, a2a-selective adenosine receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful reagent for the efficient phosphorylative conversion of alcohols into their corresponding dibenzylphosphorotriesters. It is applied as a phosphitylating agent in water-soluble phosphate prodrugs of 1-propargyl-8-styrylxanthine derivatives, a2a-selective adenosine receptor antagonists.

Solubility
Hydrolyzes in water. Soluble in chloroform and methanol.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117924-33-1
Molecular Formula C12H28NO2P
Molecular Weight (g/mol) 249.335
MDL Number MFCD00134307
InChI Key KUKSUQKELVOKBH-UHFFFAOYSA-N
Synonym di-tert-butyl n,n-diethylphosphoramidite, di-tert-butyl diethylphosphoramidite, di-t-butyl n,n-diethylphosphoramidite, phosphoramidous acid, diethyl-, bis 1,1-dimethylethyl ester, n-bis 2-methylpropan-2-yl oxy phosphanyl-n-ethylethanamine, acmc-1c42i, di-t-butyl diethylphosphoramidite, di-tert-butyl diethylamidophosphite, di-tert-butoxy diethylamino phosphine
PubChem CID 4251849
IUPAC Name N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-ethylethanamine
SMILES CCN(CC)P(OC(C)(C)C)OC(C)(C)C

Specifications

Density 0.896
Boiling Point 39°C to 41°C (0.3 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.434
Quantity 1 g
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Hydrolyzes in water. Soluble in chloroform and methanol.
Formula Weight 249.33
Percent Purity 94%
Chemical Name or Material Di-tert-butyl N,N-diethylphosphoramidite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.