missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl peroxide, 99%

Product Code. 10778152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Plastic bottle
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging unitSize
10778152 5 g Plastic bottle 5g
10367071 100 g Plastic bottle 100g
10607963 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10778152 Leverandør Thermo Scientific Acros Leverandørnr. 349930050
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format
Denne vare kan ikke returneres. Se returpolicy

CAS: 110-05-4 | C8H18O2 | 146.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 110-05-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00008803
InChI Key LSXWFXONGKSEMY-UHFFFAOYSA-N
Synonym di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d
PubChem CID 8033
SMILES CC(C)(C)OOC(C)(C)C

Tekniske data

Melting Point -30°C
Density 0.8000g/mL
Boiling Point 109°C to 110°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 0.1% max. Tert-butyl hydroperoxide (GC)
Packaging Plastic bottle
Linear Formula (CH3)3COOC(CH3)3
Refractive Index 1.3880 to 1.39
Quantity 5 g
Merck Index 15, 3508
Specific Gravity 0.8
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in most organic solvents
IUPAC Name 2-tert-butylperoxy-2-methylpropane
Viscosity 0.9 mPa.s (20°C)
Formula Weight 146.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Di-tert-butyl peroxide
Vis mere Vis mindre
compliance-icons compliance-icons
Produktidentifikator
  • Di-tert-butyl peroxide
Signalord
  • Danger
Farekategori
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Organic peroxide Type E & F
Faresætning
  • H341-Suspected of causing genetic defects.
  • H412-Harmful to aquatic life with long lasting effects.
  • H242-Heating may cause a fire.
  • H225-Highly flammable liquid and vapour.
Sikkerhedssætning
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.