Learn More
Di-tert-butyl phosphite, 95%
Beskrivelse
Di-tert-butyl phosphite is used as a solvent, as an antioxidant, and as an intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Density | 0.995 g/mL |
| Boiling Point | 66°C to 68°C (0.5 mmHg) |
| Refractive Index | 1.42 |
| Quantity | 1 g |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in organic solvents |
| Formula Weight | 194.21 |
| Percent Purity | 95% |
| Physical Form | Liquid |
| Chemical Name or Material | Di-tert-butyl phosphite |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.