missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl phosphite, 96%

Product Code. 13478658 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13478658 25 g Glass bottle 25g
13458658 1 g Glass bottle 1g
13468658 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13478658 Supplier Thermo Scientific Acros Supplier No. 449390250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13086-84-5 | C8H18O3P | 193.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13086-84-5
Molecular Formula C8H18O3P
Molecular Weight (g/mol) 193.20
MDL Number MFCD00014999,MFCD00014999
InChI Key GEBLOQXLELCEEO-UHFFFAOYSA-N
Synonym di-tert-butyl phosphite, di-tert-butylphosphite, phosphorous acid, bis 1,1-dimethylethyl ester
PubChem CID 21959072
IUPAC Name ditert-butyl phosphite
SMILES CC(C)(C)O[P+](=O)OC(C)(C)C

Tekniske data

Density 0.9950g/mL
Boiling Point 66°C to 68°C (0.5 mmHg)
Flash Point 163°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3CO]2P(O)H
Quantity 25 g
Specific Gravity 0.995
Solubility Information Solubility in water: immiscible
Formula Weight 193.2
Percent Purity 96%
Chemical Name or Material Di-tert-butyl phosphite
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Di-tert-butyl phosphite
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.