missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Phosphite, TRC

Product Code. 30378522
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378522 10 g -
30371905 5 g 5g
30378038 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378522 Supplier Toronto Research Chemicals Supplier No. TRCD42951510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13086-84-5
Molecular Formula C8H19O3P
Molecular Weight (g/mol) 194.21
Synonym Phosphonic Acid Bis(1,1-dimethylethyl) Ester, Phosphonic Acid Di-tert-butyl Ester, Di-tert-butyl Phosphonate
IUPAC Name 2-methyl-2-[(2-methylpropan-2-yl)oxyphosphonoyloxy]propane
SMILES CC(C)(C)OP(=O)OC(C)(C)C

Tekniske data

Quantity 10 g
InChI Formula InChI=1S/C8H19O3P/c1-7(2,3)10-12(9)11-8(4,5)6/h12H,1-6H3
Formula Weight 194.1072
Chemical Name or Material Di-tert-butyl Phosphite
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.