missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butylchlorophosphine, 96%

Product Code. 11335349
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335349 25 g 25g
11315349 1 g 1g
11325349 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11335349 Supplier Thermo Scientific Alfa Aesar Supplier No. B20998.14
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 13716-10-4 | C8H18ClP | 180.656 g/mol

Di-tert-butylchlorophosphine is associated with palladium(II) acetate and used in Suzuki-Miyaura cross-couplings of arylboronic acids with aryl bromides and chlorides. It serves as a ligand and used in the preparation of mono- and bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations. Further, it is used in asymmetric and transition metal coupling reactions. In addition to this, it plays an important role in the preparation of other phosphines in catalytic polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butylchlorophosphine is associated with palladium(II) acetate and used in Suzuki-Miyaura cross-couplings of arylboronic acids with aryl bromides and chlorides. It serves as a ligand and used in the preparation of mono- and bidentate phosphine ligands in metal complexes for catalytic chiral asymmetric hydrogenations. Further, it is used in asymmetric and transition metal coupling reactions. In addition to this, it plays an important role in the preparation of other phosphines in catalytic polymerization reactions.

Solubility
Miscible with terahydrofuran.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13716-10-4
Molecular Formula C8H18ClP
Molecular Weight (g/mol) 180.656
MDL Number MFCD00008815
InChI Key MCRSZLVSRGTMIH-UHFFFAOYSA-N
Synonym di-tert-butylchlorophosphine, di-tert-butylchlorophosphane, di-t-butylchlorophosphine, phosphinous chloride, bis 1,1-dimethylethyl, chlorodi-tert-butylphosphine, phosphinous chloride, di-tert-butyl, di-tert-butyl chloro phosphane, di-tert-butylphosphinous chloride, clptbu2, pubchem6477
PubChem CID 139566
IUPAC Name ditert-butyl(chloro)phosphane
SMILES CC(C)(C)P(C(C)(C)C)Cl

Specifications

Density 0.951
Boiling Point 47°C to 49°C (3 mmHg)
Flash Point 62°C (144°F)
Odor Acrid
Linear Formula [(CH3)3C]2PCl
Refractive Index 1.484
Quantity 25 g
UN Number UN3265
Sensitivity Air and moisture sensitive
Solubility Information Miscible with terahydrofuran.
Formula Weight 180.66
Percent Purity 96%
Chemical Name or Material Di-tert-butylchlorophosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.