missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butylchlorophosphine, 96%

Product Code. 10617684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10617684 100 mL Glass bottle 100mL
10093870 5 mL Glass bottle 5mL
10086383 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10617684 Supplier Thermo Scientific Acros Supplier No. 320901000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13716-10-4 | C8H18ClP | 180.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13716-10-4
Molecular Formula C8H18ClP
Molecular Weight (g/mol) 180.66
MDL Number MFCD00008815
InChI Key MCRSZLVSRGTMIH-UHFFFAOYSA-N
Synonym di-tert-butylchlorophosphine, di-tert-butylchlorophosphane, di-t-butylchlorophosphine, phosphinous chloride, bis 1,1-dimethylethyl, chlorodi-tert-butylphosphine, phosphinous chloride, di-tert-butyl, di-tert-butyl chloro phosphane, di-tert-butylphosphinous chloride, clptbu2, pubchem6477
PubChem CID 139566
IUPAC Name ditert-butyl(chloro)phosphane
SMILES CC(C)(C)P(C(C)(C)C)Cl

Specifications

Melting Point 2.0°C to 3.0°C
Color Colorless
Density 0.9500g/mL
Boiling Point 69.0°C to 70.0°C (10.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3C]2PCl
Quantity 100 mL
Specific Gravity 0.95
Formula Weight 180.66
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Di-tert-butylchlorophosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.