missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyltin dichloride, 98%, ACROS Organics™

Product Code. 10227330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10227330 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10227330 Supplier Thermo Scientific Acros Supplier No. 316740010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 19429-30-2
Melting Point 39°C to 43°C
Color White
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H18Cl2Sn
Linear Formula [(CH3)3C]2SnCl2
MDL Number MFCD00009854
Quantity 1 g
Synonym di-tert-butyldichlorostannane, di-tert-butyltin dichloride, di-t-butyltin dichloride, ditert-butyl dichloro stannane, acmc-1bpuo, di-tert-butylzinndichlorid
InChI Key PEGCFRJASNUIPX-UHFFFAOYSA-L
SMILES CC(C)(C)[Sn](C(C)(C)C)(Cl)Cl
IUPAC Name ditert-butyl(dichloro)stannane
Molecular Weight (g/mol) 303.842
PubChem CID 88053
Formula Weight 303.83
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Di-tert-butyltin dichloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.