missing translation for 'onlineSavingsMsg'
Learn More

Diacetone-D-galactose, 95%

Product Code. 11441367
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441367 25 g 25g
11431367 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441367 Supplier Thermo Scientific Alfa Aesar Supplier No. B24899.14
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4064-06-6 | C12H20O6 | 260.29 g/mol

Diacetone-D-galactose is used to produce diacetone-d-galacturonic acid and oxalic acid. It is mainly used in biochemical reaction and used as medicine intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetone-D-galactose is used to produce diacetone-d-galacturonic acid and oxalic acid. It is mainly used in biochemical reaction and used as medicine intermediate.

Solubility
Soluble in Acetone, Chloroform and Methanol.

Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4064-06-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.29
MDL Number MFCD00063225
InChI Key POORJMIIHXHXAV-UHFFFAOYNA-N
Synonym 1,2:3,4-di-o-isopropylidene-d-galactose, 1,2:3,4-di-o-isopropylidene-, a-d-galactopyranose, 1,2:3,4-bis-o-1-methylethylidene-, 1s,6r,9s-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 7.3.0.0 2 ,? dodecan-8-yl methanol
PubChem CID 70793572
IUPAC Name [(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydro-3aH-di[1,3]dioxolo[4,5-a
SMILES CC1(OC2C(OC3C(C2O1)OC(O3)(C)C)CO)C

Specifications

Density 1.143
Quantity 25 g
Solubility Information Soluble in Acetone,Chloroform and Methanol.
Formula Weight 260.29
Percent Purity 97%
Chemical Name or Material Diacetone-D-galactose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.