missing translation for 'onlineSavingsMsg'
Learn More

Diacetoxydimethylsilane, 95%, Thermo Scientific™

Product Code. 10461304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10461304 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10461304 Supplier Thermo Scientific Acros Supplier No. 426690250
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 2182-66-3
Melting Point -12.5°C
Color Colorless
Density 1.0530g/mL
Boiling Point 164°C
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C6H12O4Si
Linear Formula (CH3COO)2Si(CH3)2
Refractive Index 1.4020 to 1.4040 (20°C, 589nm)
MDL Number MFCD00039832
Quantity 25 g
Specific Gravity 1.053
Synonym dimethyldiacetoxysilane, diacetoxydimethylsilane, dimethylsilanediyl diacetate, silanediol, dimethyl-, diacetate, dimethylsilanediol, diacetate, silanediol, 1,1-dimethyl-, 1,1-diacetate, acetyloxy dimethylsilyl acetate, dimethyldiacetoxysilicane, dimethyl-silanediodiacetate, acmc-1ak12
InChI Key RQVFGTYFBUVGOP-UHFFFAOYSA-N
SMILES CC(=O)O[Si](C)(C)OC(=O)C
IUPAC Name [acetyloxy(dimethyl)silyl] acetate
Molecular Weight (g/mol) 176.24
PubChem CID 75130
Formula Weight 176.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diacetoxydimethylsilane
Show More Show Less
compliance-icons
Product Identifier
  • Diacetoxydimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.