missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Diagnostic Kit for Free Thyroxine (FT4) (MPQuanti™ only)

Product Code. 30280618 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Free Thyroxine (T4)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
30280618 Free Thyroxine (T4) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280618 Supplier MP Biomedicals Supplier No. 07FT45025
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diagnostic Kit for Free Thyroxine (FT4) is intended for in-vitro quantitative determination of free Thyroxine (FT4) in human serum or plasma.

  • Diagnostic Kit for Free Thyroxine (FT4) (Serum/Plasma) is based on fluorescence immunoassay technology.
  • The sample moves through the strip from sample pad to absorbent pad. FT4 in the sample will compete with the T4 antigen coated on the membrane.
  • The less FT4 in the sample, the more fluorescent microspheres conjugated with anti-T4 antibodies can be captured by the T4 antigen coated on the membrane (Test line).
  • The concentration of FT4 in the sample is inversely related to the intensity of the fluorescent signal captured on the T line.
  • According to the fluorescence intensity of the test and the standard curve, the concentration of FT4 in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 1 ̴ 100pmol/L, R≥0.990
  • Test results should be interpreted at 15 minutes

Specifications

Detectable Analytes Free Thyroxine (T4)
Detection Method Immunofluorescence
Sample Type Human serum, Human plasma
Type POC, Quantitative, free, T4, FT4
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.