missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Diagnostic Kit for Free Triiodothyronine (FT3) (MPQuanti™ only)

Product Code. 30280605 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Free Triiodothyronine (T3)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
30280605 Free Triiodothyronine (T3) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280605 Supplier MP Biomedicals Supplier No. 07FT35025
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diagnostic Kit for Free Triiodothyronine (FT3) is intended for in-vitro quantitative determination of free Triiodothyronine (FT3) in human serum or plasma.

  • Diagnostic Kit for Free Triiodothyronine (FT3) (Serum/Plasma) detects FT3 based on Fluorescence Immunoassay.
  • The specimen moves through the strip from sample pad to absorbent pad.
  • FT3 in the specimen will compete with the T3 antigen coated on the strip.
  • The less FT3 in the specimen, the more fluorescent microspheres conjugated with anti-T3 antibodies can be captured by the T3 antigen coated on the strip.
  • The concentration of FT3 in the sample is inversely related to the intensity of the fluorescent signal captured on the T line.
  • According to the fluorescence intensity of the test and the standard curve, the concentration of FT3 in the sample is calculated by MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 1.5-46 pmol/L (0.88-29.8 pg/mL), R≥0.990
  • Test results should be interpreted at 15 minutes

Specifications

Detectable Analytes Free Triiodothyronine (T3)
Detection Method Immunofluorescence
Sample Type Human serum, Human plasma
Type POC, Quantitative, Free, T3, FT3
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.