missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Diagnostic Kit for HbA1c (MPQuanti™ only)

Product Code. 30280620 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
HbA1c
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
30280620 HbA1c 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280620 Supplier MP Biomedicals Supplier No. 07HBA5025
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diagnostic Kit for HbA1c is intended for in-vitro quantitative determination of HbA1c in human whole blood as an aid in the diagnosis of diabetes and in identifying patients who may be at risk for developing diabetes.

  • Diagnostic Kit for HbA1c (Whole Blood) is based on fluorescence immunoassay technology.
  • The sample moves through the strip from sample pad to absorbent pad. HbA1c in the sample, attaches to the HbA1c antibody which is conjugated with fluorescent microspheres.
  • Then captured by hemoglobin (Hb) antibody coated on the nitrocellulose membrane.
  • The concentration of HbA1c in the sample correlates linearly with the fluorescence signal intensity.
  • According to the two fluorescence intensity, the MPQuanti™ Immuflorescence Analyzer calculate the value of HbA1c percentage in the sample.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 4 ̴ 14.5%, R≥0.990
  • Test results should be interpreted at 10 minutes
  • A marker of long-term metabolic control in persons with diabetes mellitus

Specifications

Detectable Analytes HbA1c
Detection Method Immunofluorescence
Sample Type Human whole blood
Type POC, Quantitative, HbA1c
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.