missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Diagnostic Kit for β-hCG (MPQuanti™ only)

Product Code. 30280609 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 Tests/Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280609 25 Tests/Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280609 Supplier MP Biomedicals Supplier No. 07HCG5025
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diagnostic Kit for β-hCG is intended for in-vitro quantitative determination of β-human chorionic gonadotropin (β-hCG) in human whole blood, serum, or plasma.

  • Diagnostic Kit for β-hCG (Whole blood/Serum/Plasma) is based on fluorescence immunoassay technology.
  • The Specimen moves from specimen pad to absorbent pad. If the specimen contains β-hCG, it attaches to the fluorescent microspheres-conjugated anti-β-hCG antibodies.
  • Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
  • The concentration of β-hCG in the specimen correlates linearly with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and standard curve, the concentration of B-hCG in the pecimen is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 2-200,000 mIU/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
  • For the diagnosis of β-human chorionic gonadotropin (β-hCG) to assess pregnancy

Specifications

Content And Storage 4°C to 30°C
Detectable Analytes beta-hCG
Sample Type Human whole blood, Human serum, Human plasma
Type POC, Quantitative, beta-hCG, pregnancy
Quantity 25 Tests/Kit
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.