missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Diagnostic Kit for Interleukin-6 (MPQuanti™ only)

Product Code. 30280617 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Interleukin-6
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
30280617 Interleukin-6 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280617 Supplier MP Biomedicals Supplier No. 07IL5025
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Diagnostic Kit for Interleukin-6 is intended for in-vitro quantitative determination of Interleukin-6 (IL-6) in human whole blood, serum, or plasma.

  • Diagnostic Kit for Interleukin-6 (Whole blood/serum/plasma) is based on fluorescence immunoassay technology.
  • The sample moves through the strip from sample pad to absorbent pad. If the specimen contains IL-6, it attaches to the fluorescent microspheres-conjugated anti-IL-6 antibodies.
  • Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
  • The concentration of IL-6 in the sample correlates linearly with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and standard curve, the concentration of IL-6 in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 3 5000 pg/mL, R≥0.990
  • Test results should be interpreted at 15 minutes
  • For diagnosis of inflammatory condition and immune status

Spezifikation

Detectable Analytes Interleukin-6
Detection Method Immunofluorescence
Sample Type Human whole blood, Human serum, Human plasma
Type POC, Quantitative, interleukin-6, IL-6
Form Ready-to-Use (RTU)
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.