missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Diagnostic Kit for Prolactin (PRL) (MPQuanti™ only)

Product Code. 30280597 Shop All MP Biomedicals Products
Change view
Click to view available options
Detectable Analytes:
Prolactin
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detectable Analytes Unit Size
30280597 Prolactin 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280597 Supplier MP Biomedicals Supplier No. 07PLAC5025
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diagnostic Kit for Prolactin (PRL) is intended for in-vitro quantitative determination of Prolactin (PRL) in human whole blood, serum, or plasma.

  • The PRL Test Cassette (Whole Blood/Serum/Plasma) is based on Fluorescence Immunoassay for the quantitative detection of Prolactin (PRL) in human whole blood, serum or plasma.
  • The sample moves through the strip from sample pad to absorbent pad. If the specimen contains PRL, it attaches to the fluorescent microspheres-conjugated anti-PRL antibodies.
  • Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
  • The concentration of PRL in the sample correlates linearly with the fluorescence signal intensity captured on the T line.
  • According to the fluorescence intensity of the test and standard curve, the concentration of PRL in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer to show PRL concentration in the sample.
  • CE marked
  • For professional in-vitro diagnostic use
  • Working only with MPQuanti™ Immunofluorescence Analyzer
  • Linear working range: 1-200 ng/mL, R≥0.990
  • Test results should be interpreted at 15 minutes

Specifications

Detectable Analytes Prolactin
Detection Method Immunofluorescence
Sample Type Human whole blood, Human serum, Human plasma
Type POC, Quantitative, prolactin, PRL
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.