missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Diagnostik™ Rack Swinging Bucket Rotor

Product Code. 11836470 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
Varies per Rack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
11836470 Varies per Rack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11836470 Supplier Thermo Scientific™ Supplier No. 75006480
  • ONLINE EXCLUSIVE PRICE
    10975.00 €
    10600.00 € / Pack of 1
    Save 375.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accommodate a range of tube sizes with a wide variety of trays by using Thermo Scientific™ Diagnostik™ Rack Swinging-Bucket Rotor.

Accommodate a range of tube sizes with a wide variety of trays by using Thermo Scientific™ Diagnostik™ Rack Swinging-Bucket Rotor, for use with Cryofuge 5500i centrifuges.

Ordering Information:

Trays for these rotors are sold separately; order rotor (75006480) which includes rotor body (70060333), rotor bucket (qty 2.) (70060335) and grease (70006692). Contact your sales representative for information.

TRUSTED_SUSTAINABILITY

Specifications

Description Diagnostik Rack Swinging Bucket Rotor
Material Stainless Steel Rotor Body, Aluminum Bucket
Type Swinging Bucket Rotor
Capacity Varies per Rack
Max. RCF 2,550 x g
Max. Speed 3,500 rpm
Radius (Metric) 186 mm
Radius (Metric) Min. 141 mm
For Use With (Equipment) Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges
Angle 90°
Min. RCF 1,929 x g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.