missing translation for 'onlineSavingsMsg'
Learn More

Dialifos, TRC

Product Code. 30366767
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366767 250 mg 250mg
30360202 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30366767 Supplier Toronto Research Chemicals Supplier No. TRCD416123250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10311-84-9
Molecular Formula C14 H17 Cl N O4 P S2
Molecular Weight (g/mol) 393.85
Synonym Phosphorodithioic acid, O,O-diethyl ester, S-ester with N-(2-chloro-1-mercaptoethyl)phthalimide (7CI,8CI), Phthalimide, N-(2-chloro-1-mercaptoethyl)-, S-ester with O,O-diethyl phosphorodithioate (8CI), Dialifor, Dialifos, Dialiphor, Dialiphos, Hercules 14503, O,O-Diethyl S-(2-chloro-1-phthalimidoethyl) phosphorodithioate, S-(2-Chloro-1-phthalimidoethyl) O,O-diethyl phosphorodithioate, Torak
IUPAC Name 2-(2-chloro-1-diethoxyphosphinothioylsulfanylethyl)isoindole-1,3-dione
SMILES CCOP(=S)(OCC)SC(CCl)N1C(=O)c2ccccc2C1=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H17ClNO4PS2/c1-3-19-21(22,20-4-2)23-12(9-15)16-13(17)10-7-5-6-8-11(10)14(16)18/h5-8,12H,3-4,9H2,1-2H3
Formula Weight 393.0025
Chemical Name or Material Dialifos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.