missing translation for 'onlineSavingsMsg'
Learn More

Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide

Product Code. 11387556
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387556 25 g 25g
11377556 5 g 5g
11397556 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387556 Supplier Thermo Scientific Alfa Aesar Supplier No. L11106.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diallyl disulfide, 80%, C6H10S2, CAS Number-2179-57-9, diallyldisulfide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene, 2-propenyl disulphide, diallyl disulfide, allyl disulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 100g | CAS: 2179-57-9 | C6H10S2 | 146.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2179-57-9
Molecular Formula C6H10S2
Molecular Weight (g/mol) 146.27
MDL Number MFCD00008656
InChI Key PFRGXCVKLLPLIP-UHFFFAOYSA-N
Synonym diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene
PubChem CID 16590
ChEBI CHEBI:4488
IUPAC Name 3-(prop-2-enyldisulfanyl)prop-1-ene
SMILES C=CCSSCC=C

Specifications

Density 1.008
Boiling Point 180°C to 190°C
Flash Point 51°C (123°F)
Odor Garlic-like
Linear Formula CH2=CHCH2S-SCH2CH=CH2
Refractive Index 1.541
Quantity 25 g
UN Number UN1993
Beilstein 1699241
Formula Weight 146.27
Percent Purity 80%
Grade Technical
Assay remainder mainly diallyl sulfide and diallyl trisulfide
Chemical Name or Material Diallyl disulfide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diallyl disulfide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Diallyl disulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.