missing translation for 'onlineSavingsMsg'
Learn More

Diallyl Phthalate-d4, TRC

Product Code. 30355458
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30355458 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30355458 Supplier Toronto Research Chemicals Supplier No. TRCD41605710MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-allyl ester D4
CAS 2514944-45-5
Molecular Formula C14 D4 H10 O4
Quantity 10 mg
Synonym Phthalic Acid, Bis-allyl Ester D4 (phenyl D4), Diallyl Phthalate-d4, Diallyl Phthalate-3,4,5,6-d4
InChI Formula InChI=1S/C14H14O4/c1-3-9-17-13(15)11-7-5-6-8-12(11)14(16)18-10-4-2/h3-8H,1-2,9-10H2/i5D,6D,7D,8D
SMILES [2H]c1c([2H])c([2H])c(C(=O)OCC=C)c(C(=O)OCC=C)c1[2H]
IUPAC Name bis(prop-2-enyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 250.2832
Formula Weight 250.1143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.