Learn More
Gilson™ DIAMOND™ Steril-Pack™ Sterile Pipet Tips
Product Code. 10307712 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
100 to 1000 μL
2 to 200 μL
Unit Size:
Pack of 400
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10307712 2 to 200 μL Pack of 400
10123002 100 to 1000 μL Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10307712 Supplier Gilson™ Supplier No. F161330
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Traceable from the mould to the laboratory with batch number on each pack and rack. Diamond™ Steril-Pack™ Sterile Pipet Tips can be purchased sterilized or for autoclaving.
  • Tips comply with the CE IVD directive
  • Color coded packaging for volume recognition at a glance
  • Sterilized tips are free of detectable DNA, RNA, DNase, RNase, ATP, pyrogens and trace metals
  • Pre-sterilized tips are wrapped in medical device quality plastic
  • Gamma irradiated
  • Non-sterile tips can be autoclaved at 121°C for 20 minutes at 1 bar (moist heat)
TRUSTED_SUSTAINABILITY
Specifications
Description Pipette tip
Volume (Metric) 2 to 200 μL
Sterility Sterile
Autoclavable Autoclavable
Model D200
Precision 0.006 to 0.008μL
Accuracy -0.075 to -0.05μL
Length (English) 2 in.
Length (Metric) 5.1 cm
Certifications/Compliance CE, IVD
Clean Claims DNA, RNA, DNase, RNase, ATP, Pyrogen-free and trace Metals free
For Use With (Application) Liquid handling, medical, pharmaceutical and food industries, in various fields and applications such as PCR, molecular biology or cell technology
Tip Style Universal
Packaging Box
Quantity 400 Pk.
Product Type Pipet Tips
For Use With (Equipment) Gilson Pipetman F20L, Gilson Pipetman F25L, Gilson Pipetman F50L, Gilson Pipetman F100L, Gilson Pipetman F200L, Gilson Pipetman P200L, Gilson Pipetman P20M, Gilson Pipetman P200M, Gilson Pipetman
Graduated Yes
Product Line Diamond Sterlipack
Material Polypropylene