missing translation for 'onlineSavingsMsg'
Learn More

Diatomaceous earth, Flux-calcined, TRC

Product Code. 30349967
Change view
Click to view available options
Quantity:
1 Kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30349967 1 Kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349967 Supplier Toronto Research Chemicals Supplier No. TRCD4168211KG
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diatomaceous earth, Flux-calcined
CAS 68855-54-9
Molecular Formula SiO2
Quantity 1 Kg
Synonym Filter Agent, Diatomaceous Earth, Flux-calcined, Celatom FW-14, Celite™ 545, Particle Size 0.02-0.1 mm, Kieselguhr, soda ash flux-calcined
SMILES O=[Si]=O
Molecular Weight (g/mol) 60.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.