missing translation for 'onlineSavingsMsg'
Learn More

Diatomaceous Earth, J.T. Baker™

Product Code. 15527944 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15527944 500 g Wide Mouth Amber Glass Bottle 500g
15538774 1 kg Poly Pail 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15527944 Supplier Avantor J.T.Baker Supplier No. 193901

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Packaging Wide Mouth Amber Glass Bottle
Particle Size 33 Micron
Specific Gravity 1L = 2.30kg
Quantity 500 g
Chemical Name or Material Diatomaceous Earth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.