missing translation for 'onlineSavingsMsg'
Learn More

Diazolidinyl urea, 95%

Product Code. 10352020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10352020 25 g Glass bottle 25g
10647040 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10352020 Supplier Thermo Scientific Acros Supplier No. 428580250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78491-02-8 | C8H14N4O7 | 278.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78491-02-8
Molecular Formula C8H14N4O7
Molecular Weight (g/mol) 278.22
MDL Number MFCD03547942
InChI Key SOROIESOUPGGFO-UHFFFAOYSA-N
Synonym diazolidinylurea, diazolidinyl urea, 1-1,3-bis hydroxymethyl-2,5-dioxoimidazolidin-4-yl-1,3-bis hydroxymethyl urea, germall ii, germall 11, imidazolidinyl urea 11, n,n'-bis hydroxymethyl urea, dsstox_cid_9559, dsstox_rid_78776
PubChem CID 62277
IUPAC Name 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea
SMILES C(NC(=O)N(CO)C1C(=O)N(C(=O)N1CO)CO)O

Specifications

Melting Point 227.0°C to 228.0°C
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Merck Index 15,3003
Solubility Information Solubility in water: soluble
Formula Weight 278.22
Percent Purity 95%
Chemical Name or Material Diazolidinyl urea
compliance-icons
Product Identifier
  • Diazolidinyl urea
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.