missing translation for 'onlineSavingsMsg'
Learn More

Dibenz[a,h]anthracene, TRC

Product Code. 18081340
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18081340 100 mg 100mg
18041240 1 g 1g
18081240 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18081340 Supplier Toronto Research Chemicals Supplier No. TRCD417010100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-70-3
Molecular Formula C22 H14
Molecular Weight (g/mol) 278.35
Synonym 1,2:5,6-Benzanthracene, 1,2:5,6-Dibenz[a]anthracene, 1,2:5,6-Dibenzanthracen, 1,2:5,6-Dibenzanthracene, 1,2:5,6-Dibenzoanthracene, DBA, Dibenzo[a,h]anthracene, NSC 22433
IUPAC Name naphtho[1,2-b]phenanthrene
SMILES c1ccc2c(c1)ccc3cc4c(ccc5ccccc45)cc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H14/c1-3-7-19-15(5-1)9-11-17-14-22-18(13-21(17)19)12-10-16-6-2-4-8-20(16)22/h1-14H
Formula Weight 278.1096
Percent Purity >95%
Chemical Name or Material Dibenzo(a,h)anthracene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.