missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo-18-crown-6, TRC

Product Code. 30376797
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
missing translation for 'unitSize':
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30376797 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30376797 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD53392550MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzo-18-crown-6
CAS 14187-32-7
Molecular Formula C20H24O6
Quantity 50 mg
InChI Formula InChI=1S/C20H24O6/c1-2-6-18-17(5-1)23-13-9-21-11-15-25-19-7-3-4-8-20(19)26-16-12-22-10-14-24-18/h1-8H,9-16H2
SMILES C1COc2ccccc2OCCOCCOc3ccccc3OCCO1
Molecular Weight (g/mol) 360.4
Formula Weight 360.1573
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.