missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[b,d]thiophen-2-ylboronic acid, 97%, Thermo Scientific™

Product Code. 10462603
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10462603 250 mg Amber Glass Bottle 250mg
10278522 1 g Amber Glass Bottle 1g
10336854 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10462603 Supplier Thermo Scientific Maybridge Supplier No. MO01033CB
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 668983-97-9
Molecular Formula C12H9BO2S
Molecular Weight (g/mol) 228.072
MDL Number MFCD01318982
InChI Key CSLSCVHILGCSTE-UHFFFAOYSA-N
Synonym dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l
PubChem CID 2794660
IUPAC Name dibenzothiophen-2-ylboronic acid
SMILES B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 228.08
Percent Purity 95%
Chemical Name or Material Dibenzo[b, d]thiophen-2-ylboronic acid, May contain varying amounts of anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.