missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[b,d]thiophene-4-boronic acid, TRC

Product Code. 30367611
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367611 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367611 Supplier Toronto Research Chemicals Supplier No. TRCD417313100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzo[b,d]thiophene-4-boronic acid
CAS 108847-20-7
Molecular Formula C12H9BO2S
Quantity 100 mg
InChI Formula InChI=1S/C12H9BO2S/c14-13(15)10-6-3-5-9-8-4-1-2-7-11(8)16-12(9)10/h1-7,14-15H
SMILES OB(O)c1cccc2c3ccccc3sc12
IUPAC Name dibenzothiophen-4-ylboronic acid
Molecular Weight (g/mol) 228.08
Formula Weight 228.0416
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.