missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[b,f][1,4]thiazepine-11-[10H]one, TRC

Product Code. 30005502
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005502 5 g 5g
17970785 1 g 1g
17901025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005502 Supplier Toronto Research Chemicals Supplier No. TRCD4169535G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3159-07-7
Molecular Formula C13 H9 N O S
Molecular Weight (g/mol) 227.28
Synonym Dibenzo[b,f][1,4]thiazepin-11(10H)-one, Quetiapine Fumarate Imp. G (EP)
IUPAC Name 5H-benzo[b][1,4]benzothiazepin-6-one
SMILES O=C1Nc2ccccc2Sc3ccccc13

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H9NOS/c15-13-9-5-1-3-7-11(9)16-12-8-4-2-6-10(12)14-13/h1-8H,(H,14,15)
Formula Weight 227.0405
Percent Purity >95%
Chemical Name or Material Dibenzo[b,f][1,4]thiazepin-11(10H)-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.