missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran, 98%

Product Code. 11384519
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384519 25 g 25g
11394519 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384519 Supplier Thermo Scientific Alfa Aesar Supplier No. A16521.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 132-64-9 | C12H8O | 168.195 g/mol

Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.

Solubility
Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-64-9
Molecular Formula C12H8O
Molecular Weight (g/mol) 168.195
MDL Number MFCD00004968
InChI Key TXCDCPKCNAJMEE-UHFFFAOYSA-N
Synonym dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide
PubChem CID 568
ChEBI CHEBI:28145
IUPAC Name dibenzofuran
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3O2

Specifications

Melting Point 78°C to 83°C
Density 1.3
Boiling Point 287°C to 289°C
Flash Point 130°C (266°F)
Quantity 25 g
UN Number UN3077
Beilstein 121100
Solubility Information Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents.
Formula Weight 168.2
Percent Purity 98%
Chemical Name or Material Dibenzofuran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.