missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran, TRC

Product Code. 30357570
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357570 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357570 Supplier Toronto Research Chemicals Supplier No. TRCD41718550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzofuran
CAS 132-64-9
Molecular Formula C12 H8 O
Quantity 50 g
Synonym Dibenzofuran, 2,2'-Biphenylene oxide, 2,2'-Biphenylylene oxide, Dibenzo[b,d]furan, Diphenylene oxide, NSC 1245, [1,1'-Biphenyl]-2,2'-diyl oxide
InChI Formula InChI=1S/C12H8O/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H
SMILES o1c2ccccc2c3ccccc13
IUPAC Name dibenzofuran
Molecular Weight (g/mol) 168.19
Formula Weight 168.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.