missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene-2-boronic acid, TRC

Product Code. 30349852
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30349852 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349852 Supplier Toronto Research Chemicals Supplier No. TRCD417373250MG
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzothiophene-2-boronic acid
CAS 668983-97-9
Molecular Formula C7 H3 Cl F2 O
Quantity 250 mg
Synonym B-2-Dibenzothienylboronic Acid, (Dibenzothiophen-2-yl)boronic Acid, 2-Dibenzothienylboronic Acid, Dibenzo[b,d]thiophen-2-ylboronic Acid, Dibenzothiophene-2-boronic Acid
InChI Formula InChI=1S/C7H3ClF2O/c8-5-2-6(9)4(3-11)1-7(5)10/h1-3H
SMILES Fc1cc(C=O)c(F)cc1Cl
IUPAC Name 4-chloro-2,5-difluorobenzaldehyde
Molecular Weight (g/mol) 176.548
Formula Weight 175.984
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.