missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene-4-boronic acid, 95%

Product Code. 11319815
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319815 5 g 5g
11309815 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319815 Supplier Thermo Scientific Alfa Aesar Supplier No. L19831.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108847-20-7 | C12H9BO2S | 228.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108847-20-7
Molecular Formula C12H9BO2S
Molecular Weight (g/mol) 228.07
MDL Number MFCD01318182
InChI Key GOXNHPQCCUVWRO-UHFFFAOYSA-N
Synonym dibenzothiophene-4-boronic acid, 4-dibenzothiopheneboronic acid, dibenzo b,d thiophen-4-ylboronic acid, 4-dibenzothienylboronic acid, dibenzo b,d thiophene-4-boronic acid, boronic acid, 4-dibenzothienyl, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid, pubchem7386, 4-dibenzothiopheneboronicacid
PubChem CID 2734329
IUPAC Name dibenzothiophen-4-ylboronic acid
SMILES OB(O)C1=CC=CC2=C1SC1=C2C=CC=C1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 197863
Formula Weight 228.08
Percent Purity 95%
Chemical Name or Material Dibenzothiophene-4-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.