missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene, 98%

Product Code. 15189153 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15189153 1 kg Plastic Bottle 1kg
10010800 25 g Plastic Bottle 25g
10696102 100 g Plastic Bottle 100g
10003903 250 g Plastic Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15189153 Supplier Thermo Scientific Acros Supplier No. 112540010
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 132-65-0 | C12H8S | 184.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-65-0
Molecular Formula C12H8S
Molecular Weight (g/mol) 184.26
MDL Number MFCD00004969
InChI Key IYYZUPMFVPLQIF-UHFFFAOYSA-N
Synonym diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene
PubChem CID 3023
ChEBI CHEBI:23681
IUPAC Name dibenzothiophene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3S2

Specifications

Melting Point 97°C to 100°C
Color Gray-Green to White
Boiling Point 332.0°C to 333.0°C
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic Bottle
Quantity 1 kg
Beilstein 17,72
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,benzene,chloroform
Formula Weight 184.26
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Dibenzothiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.