missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-D-tartaric acid, 98+%

Product Code. 10033390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
Product Code. Quantity Packaging unitSize
10033390 25 g Plastic bottle 25g
10474195 100 g Plastic bottle 100g
10484915 500 g Plastic bottle 500g
3 options
This item is not returnable. View return policy

Product Code. 10033390

Brand: Thermo Scientific Acros 302510250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17026-42-5 | C18H14O8 | 358.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17026-42-5
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00063222
InChI Key YONLFQNRGZXBBF-ZCWZLOQUNA-N
Synonym dibenzoyl-d-tartaric acid, +-dibenzoyl-d-tartaric acid, 2s,3s-2,3-bis benzoyloxy succinic acid, d-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2s,3s, +-o,o-dibenzoyltartaric acid, 2s,3s-+-dibenzoyltartaric acid, +-dibenzoyltartaric acid, +-dibenzoyl tartaric acid
PubChem CID 1550213
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Plastic bottle
Quantity 25 g
Specific Rotation Condition +120° (20°C c=5,CH3OH)
Specific Rotation + 120
Solubility Information Solubility in water: insoluble
IUPAC Name (2S,3S)-2,3-dibenzoyloxybutanedioic acid
Formula Weight 358.31
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material (+)-Dibenzoyl-D-tartaric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.