missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-D-tartaric acid, anhydrous, 99%

Product Code. 11496456
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496456 50 g 50g
11486456 10 g 10g
11406466 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496456 Supplier Thermo Scientific Alfa Aesar Supplier No. B24754.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17026-42-5 | C18H14O8 | 358.30 g/mol

For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17026-42-5
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00063222
InChI Key YONLFQNRGZXBBF-ZCWZLOQUNA-N
Synonym dibenzoyl-d-tartaric acid, +-dibenzoyl-d-tartaric acid, 2s,3s-2,3-bis benzoyloxy succinic acid, d-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2s,3s, +-o,o-dibenzoyltartaric acid, 2s,3s-+-dibenzoyltartaric acid, +-dibenzoyltartaric acid, +-dibenzoyl tartaric acid
PubChem CID 1550213
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 154°C to 156°C
Beilstein 2227343
Solubility Information Soluble in ethanol. Insoluble in water.
IUPAC Name (2S,3S)-2,3-bis(benzoyloxy)butanedioic acid
Formula Weight 358.31
Percent Purity 99%
Quantity 50 g
Optical Rotation +120° (c=5 in Methanol)
Chemical Name or Material (+)-Dibenzoyl-D-tartaric acid, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.