missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-D-tartaric acid monohydrate, 99%

Product Code. 11437827
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437827 100 g 100g
11427827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11437827 Supplier Thermo Scientific Alfa Aesar Supplier No. A16503.22
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80822-15-7 | C18H16O9 | 376.32 g/mol

(+)-Dibenzoyl-D-tartaric acid monohydrate is used as a chiral separation and racemization agent for amines. It is also used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Dibenzoyl-D-tartaric acid monohydrate is used as a chiral separation and racemization agent for amines. It is also used as a pharmaceutical raw material.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80822-15-7
Molecular Formula C18H16O9
Molecular Weight (g/mol) 376.32
MDL Number MFCD00150722
InChI Key DXDIHODZARUBLA-DZWQFRRJNA-N
Synonym 2s,3s-2,3-bis benzoyloxy succinic acid hydrate, +-dibenzoyl-d-tartaric acid monohydrate, d-dbta, dibenzoyl-d-+-tartaric acid monohydrate, o,o'-dibenzoyl d-tartaric acid monohydrate, +-2,3-dibenzoyl-d-tartaric acid monohydrate, +-dibenzoyl-d-tartaric acid, 2s,3s-+-dibenzoyl-d-tartaric acid monohydrate, 2s,3s-+-o,o'-dibenzoyltartaric acid monohydrate, butanedioic acid, 2,3-bis benzoyloxy-, monohydrate, 2s,3s
PubChem CID 11667949
SMILES O.OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 87°C to 91°C
Quantity 100 g
Optical Rotation +109° (c=2 in Ethanol)
Beilstein 6121616
Solubility Information Soluble in methanol. Slightly soluble in water.
IUPAC Name (2S,3S)-2,3-bis(benzoyloxy)butanedioic acid hydrate
Formula Weight 376.33 (358.31 Anhydrous)
Percent Purity 99%
Chemical Name or Material (+)-Dibenzoyl-D-tartaric acid monohydrate
compliance-icons
Product Identifier
  • (+)-Dibenzoyl-D-tartaric acid, monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (+)-Dibenzoyl-D-tartaric acid, monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.