missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric acid,98%

Product Code. 11494260
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494260 25 g 25g
11413257 100 g 100g
11423257 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494260 Supplier Thermo Scientific Alfa Aesar Supplier No. A16181.14
 Restricted Item
Estimated Shipment: 14-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 2743-38-6 | C18H14O8 | 358.30 g/mol

(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 2743-38-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00003074
InChI Key YONLFQNRGZXBBF-ZIAGYGMSSA-N
Synonym dibenzoyl-l-tartaric acid, 2r,3r-2,3-bis benzoyloxy succinic acid, dibenzoyltartaric acid, --dibenzoyl-l-tartaric acid, dibenzoyl-l-tartaricacid, l-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r, unii-3q14b666vm, 2,3-bis benzoyloxy tartaric acid, o,o-dibenzoyl-+-tartaric acid
PubChem CID 75969
SMILES OC(=O)[C@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Spezifikation

Melting Point 154°C to 156°C
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
IUPAC Name (2R,3R)-2,3-dibenzoyloxybutanedioic acid
Formula Weight 358.31
Percent Purity 99%
Chemical Name or Material (-)-Dibenzoyl-L-tartaric acid, anhydrous

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.